Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033155.1 | NP_001028327.1 | 8 | 1475 | ||
GenBank mRNA | AK017735.1 | - | 8 | 1336 | ||
AK135329.1 | - | 1 | 3326 | |||
BC117717.1 | - | 8 | 1276 | 40088109 | ||
BC117718.1 | - | 8 | 1276 | 40088110 |