Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042653.1 | NP_001036118.1 | 6 | 821 | ||
GenBank mRNA | AK017825.1 | - | 6 | 786 | ||
AK045323.1 | - | 5 | 1024 | |||
BC115883.1 | - | 6 | 777 | 40088361 | ||
BC116660.1 | - | 6 | 778 | 40088363 |