Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oip5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042653.1 | NP_001036118.1 | 2 | 385 | ||
GenBank mRNA | AK045323.1 | - | 1 | 588 | ||
BC115883.1 | - | 2 | 341 | 40088361 | ||
BC116660.1 | - | 2 | 342 | 40088363 | ||
CJ046601.1 | - | 2 | 386 |