Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027558.1 | NP_081834.1 | 3 | 1813 | ||
GenBank mRNA | AK014543.1 | - | 3 | 1712 | ||
AK140064.1 | - | 3 | 1793 | |||
AK165560.1 | - | 1 | 996 | |||
AK184626.1 | - | 1 | 384 | |||
AX775753.1 | - | 3 | 1769 | |||
BC044759.2 | - | 3 | 1754 |