Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027558.1 | NP_081834.1 | 3 | 708 | ||
GenBank mRNA | AK014543.1 | - | 3 | 607 | ||
AK140064.1 | - | 3 | 688 | |||
AK184727.1 | - | 2 | 258 | |||
AX775753.1 | - | 3 | 664 | |||
BC044759.2 | - | 3 | 649 |