Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027630.1 | NP_081906.1 | 4 | 1079 | ||
GenBank mRNA | AK016452.1 | - | 4 | 1080 |