Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204223.1 | NP_001191152.1 | 4 | 302 | ||
NM_027709.2 | NP_081985.1 | 3 | 216 | |||
GenBank mRNA | AK016908.1 | - | 3 | 217 | ||
BC117712.2 | - | 4 | 262 | 40088033 |