Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081224.2 | NP_001074693.1 | 2 | 722 | ||
GenBank mRNA | AK017282.1 | - | 1 | 536 | ||
AK042678.1 | - | 884 | ||||
AK045646.1 | - | 1 | 676 | |||
AK046456.1 | - | 2 | 887 | |||
AK081911.1 | - | 698 | ||||
BC132481.1 | - | 2 | 722 | 40130758 |