Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027941.1 | NP_082217.1 | 8, 9 | 929 | ||
GenBank mRNA | AK005720.1 | - | 8, 9 | 981 | ||
BC100519.1 | - | 8, 9 | 929 | 30919449 | ||
BC116419.1 | - | 7, 8 | 762 | |||
BC116420.1 | - | 7, 8 | 769 | 40103399 | ||
JF934738.1 | - | 8, 9 | 880 |