Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam217a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027967.1 | NP_082243.1 | 8 | 1205 | ||
GenBank mRNA | AK006388.1 | - | 7 | 622 | ||
AK139828.1 | - | 8 | 1209 |