Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289828.1 | NP_001276757.1 | 1 | 32 | ||
NM_001289830.1 | NP_001276759.1 | 1 | 32 | |||
NM_028016.3 | NP_082292.1 | 1 | 32 | |||
GenBank mRNA | AY455285.1 | - | 1 | 32 | ||
BC132017.1 | - | 1 | 29 | 40130294 | ||
BC137873.1 | - | 1 | 29 | 8860892 | ||
BC144995.1 | - | 1 | 29 | 9053543 | ||
BC144996.1 | - | 1 | 29 | 9053544 |