Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028070.1 | NP_082346.1 | 3 | 1227 | ||
GenBank mRNA | AK008083.1 | - | 3 | 1289 | ||
BC022729.1 | - | 3 | 1227 | 4505097 |