Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sapcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081085.2 | NP_001074554.1 | 7 | 2862 | ||
NM_001290407.1 | NP_001277336.1 | 6 | 2760 | |||
GenBank mRNA | AK166625.1 | - | 7 | 2537 |