Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028093.1 | NP_082369.1 | 10 | 1586 | ||
GenBank mRNA | AK008590.1 | - | 6 | 1108 | ||
AY364442.1 | - | 10 | 1448 | |||
BC031143.1 | - | 10 | 1582 | 4973216 |