Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 3, 4 | 847 | ||
NM_001320132.1 | NP_001307061.1 | 3, 4 | 944 | |||
NM_001320136.1 | NP_001307065.1 | 3, 4 | 825 | |||
NM_004402.3 | NP_004393.1 | 3, 4 | 775 | |||
NR_135150.1 | 3, 4 | 775 | ||||
NR_135151.1 | 4, 5 | 862 | ||||
NR_135152.1 | 3, 4 | 797 | ||||
XM_017000499.1 | XP_016855988.1 | 3, 4 | 827 | |||
XM_017000500.1 | XP_016855989.1 | 3, 4 | 582 | |||
GenBank mRNA | AB013918.1 | - | 3, 4 | 438 | ||
AB028911.1 | - | 3, 4 | 569 | |||
AB028912.1 | - | 3, 4 | 738 | |||
AB028913.1 | - | 4, 5 | 656 | |||
AF039210.1 | - | 3, 4 | 438 | |||
AF064019.1 | - | 3, 4 | 569 | |||
AF409060.1 | - | 5, 6 | 449 | |||
AF409061.1 | - | 3, 4 | 464 | |||
AF409062.1 | - | 3, 4 | 492 | |||
AK290877.1 | - | 3, 4 | 566 | |||
AK303065.1 | - | 3, 4 | 654 | |||
BC048797.1 | - | 3, 4 | 761 | 5204996 |