Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028200.1 | NP_082476.1 | 10 | 1386 | ||
GenBank mRNA | AK005046.2 | - | 10 | 1338 | ||
AK010402.1 | - | 10 | 1319 | |||
AK028751.1 | - | 10 | 1329 | |||
AK032561.1 | - | 9 | 1843 | |||
AK151498.1 | - | 10 | 1392 | |||
AK154409.1 | - | 10 | 1304 | |||
AK155433.1 | - | 10 | 1304 | |||
AK187838.1 | - | 1 | 335 | |||
AY751521.1 | - | 10 | 1559 | |||
BC021813.1 | - | 1 | 189 | |||
BC057621.1 | - | 10 | 1387 | 6402384 |