Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081390.1 | NP_001074859.1 | 2 | 646 | ||
NM_001293772.1 | NP_001280701.1 | 2 | 650 | |||
GenBank mRNA | AK052157.1 | - | 649 | |||
AK052489.1 | - | 2 | 627 |