Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081381.1 | NP_001074850.1 | 7 | 1258 | ||
GenBank mRNA | AK011270.1 | - | 8 | 1414 | ||
AK075986.1 | - | 7 | 1255 | |||
BC048732.1 | - | 7 | 1233 | 6512688 | ||
BM943556.1 | - | 1 | 154 |