Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081349.2 | NP_001074818.2 | 15 | 2522 | ||
NM_001083809.1 | NP_001077278.1 | 14 | 2461 | |||
NM_024497.1 | NP_078773.1 | 15 | 2420 | |||
GenBank mRNA | AB103034.1 | - | 15 | 2491 | ||
AK031215.1 | - | 14 | 2464 | |||
CJ307166.1 | - | 1 | 297 |