Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289591.1 | NP_001276520.1 | 1 | 108 | ||
NM_001289592.1 | NP_001276521.1 | 1 | 108 | |||
NM_028261.1 | NP_082537.1 | 1 | 108 | |||
GenBank mRNA | AK012006.1 | - | 1 | 55 | ||
AK089405.1 | - | 1 | 86 | |||
AK158458.1 | - | 1 | 17 | |||
AK170724.1 | - | 1 | 86 | |||
AK171065.1 | - | 1 | 72 | |||
AK171203.1 | - | 1 | 72 | |||
AK171612.1 | - | 1 | 39 | |||
BB871885.1 | - | 1 | 110 | |||
BB872463.1 | - | 1 | 110 | |||
BC046640.1 | - | 1 | 33 | 5067343 |