Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160326.1 | NP_001153798.1 | 3 | 512 | ||
GenBank mRNA | AK012850.1 | - | 2 | 455 | ||
AY425619.1 | - | 3 | 497 | |||
BC100483.1 | - | 5 | 616 | |||
BU057067.1 | - | 3 | 512 | |||
CB057846.1 | - | 4 | 453 | |||
CF728547.1 | - | 2 | 271 |