Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028326.1 | NP_082602.1 | 15 | 2611 | ||
GenBank mRNA | AK012894.1 | - | 1 | 350 | ||
AK173309.1 | - | 1 | 858 |