Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fance | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163819.1 | NP_001157291.1 | 2 | 429 | ||
NM_001163820.1 | NP_001157292.1 | 2 | 429 | |||
GenBank mRNA | AK013325.1 | - | 2 | 393 | ||
AK135017.1 | - | 3 | 333 | |||
AK172283.1 | - | 2 | 413 | |||
BC016538.1 | - | 2 | 387 | |||
BC039987.1 | - | 2 | 392 | |||
BY740269.1 | - | 2 | 413 |