Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278415.1 | NP_001265344.1 | 9 | 1065 | ||
NM_028388.3 | NP_082664.1 | 8 | 968 | |||
GenBank mRNA | AK146998.1 | - | 8 | 902 | ||
AK166539.1 | - | 8 | 929 | |||
AK169143.1 | - | 8 | 939 |