Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081435.1 | NP_001074904.1 | 2 | 400 | ||
GenBank mRNA | BC158049.1 | - | 2 | 335 | 9007353 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX529625.1 | - | 225 | Chr. - - [-] on Build |