Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2n2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028420.2 | NP_082696.1 | 2 | 1205 | ||
GenBank mRNA | AK013788.1 | - | 2 | 1151 |