Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110079O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028473.1 | NP_082749.1 | 3 | 452 | ||
GenBank mRNA | AK014261.1 | - | 3 | 446 | ||
BC115669.1 | - | 3 | 450 | 40104070 | ||
BC115670.1 | - | 3 | 452 | 40104071 | ||
KM657818.1 | - | 3 | 481 |