Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028593.2 | NP_082869.2 | 4 | 2093 | ||
GenBank mRNA | AK029112.1 | - | 4 | 2096 |