Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019311.1 | NP_062184.1 | 2 | 393 | ||
GenBank mRNA | U55192.1 | - | 2 | 393 |