Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004418.3 | NP_004409.1 | 3 | 805 | ||
XM_017003546.1 | XP_016859035.1 | 2 | 917 | |||
GenBank mRNA | AF331843.1 | - | 1 | 449 | ||
BC007771.2 | - | 3 | 781 | 4297852 | ||
L11329.1 | - | 3 | 804 |