Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mafb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019316.1 | NP_062189.1 | 1 | 927 | ||
GenBank mRNA | U56241.1 | - | 1 | 927 |