Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267872.1 | NP_001254801.1 | 6 | 1120 | ||
NM_001285936.1 | NP_001272865.1 | 7 | 1254 | |||
NM_001285937.1 | NP_001272866.1 | 6 | 1197 | |||
NM_028673.1 | NP_082949.1 | 7 | 1170 | |||
GenBank mRNA | AB777269.1 | - | 6 | 1197 | ||
AB777270.1 | - | 7 | 1254 | |||
AK132808.1 | - | 6 | 1121 |