Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028687.1 | NP_082963.1 | 4 | 441 | ||
GenBank mRNA | AK015067.1 | - | 4 | 430 |