Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 10, 11 | 2069 | ||
GenBank mRNA | AB093256.1 | - | 8, 9 | 1616 | ||
AK036540.1 | - | 10, 11 | 2053 | |||
AK145708.1 | - | 10, 11 | 2111 | |||
AK150045.1 | - | 10, 11 | 2029 | |||
AK154146.1 | - | 10, 11 | 2058 | |||
AK154925.1 | - | 10, 11 | 2070 | |||
AK157310.1 | - | 10, 11 | 2079 | |||
AK198311.1 | - | 1, 2 | 280 | |||
BC060069.1 | - | 5, 6 | 1181 | |||
BC094446.1 | - | 1, 2 | 649 |