Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 23 | 3687 | ||
GenBank mRNA | AB093256.1 | - | 20 | 3346 | ||
AK004841.1 | - | 11 | 1235 | |||
AK036540.1 | - | 23 | 3671 | |||
AK037082.1 | - | 8 | 767 | |||
AK145708.1 | - | 22 | 3576 | |||
AK150045.1 | - | 23 | 3647 | |||
AK154146.1 | - | 23 | 3676 | |||
AK154925.1 | - | 23 | 3688 | |||
AK157310.1 | - | 23 | 3697 | |||
BC043323.1 | - | 5 | 528 | |||
BC060069.1 | - | 18 | 2799 | |||
BC061500.1 | - | 1 | 17 | |||
BC072573.1 | - | 21 | 4386 | 5705116 | ||
BC094446.1 | - | 14 | 2267 |