Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080981.1 | NP_001074450.1 | 8 | 866 | ||
GenBank mRNA | AK014820.1 | - | 6 | 1624 | ||
BC049928.1 | - | 6 | 1596 |