Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127339.1 | NP_001120811.1 | 6, 7 | 1037 | ||
NM_001127341.1 | NP_001120813.1 | 6, 7 | 1037 | |||
NM_001127342.1 | NP_001120814.1 | 4, 5 | 608 | |||
NM_001127343.1 | NP_001120815.1 | 4, 5 | 608 | |||
NM_001127344.1 | NP_001120816.1 | 4, 5 | 608 | |||
NM_019221.3 | NP_062094.1 | 6, 7 | 1037 | |||
XM_006248500.2 | XP_006248562.1 | 6, 7 | 1118 | |||
XM_008768791.1 | XP_008767013.1 | 7, 8 | 967 | |||
GenBank mRNA | AJ277446.1 | - | 5, 6 | 839 | ||
AJ277447.1 | - | 4, 5 | 608 | |||
AJ277448.1 | - | 5, 6 | 839 | |||
AJ277449.1 | - | 6, 7 | 890 | |||
AJ277450.1 | - | 4, 5 | 608 | |||
AJ277451.1 | - | 6, 7 | 890 | |||
AJ277452.1 | - | 5, 6 | 839 | |||
AJ277453.1 | - | 4, 5 | 608 | |||
Y10258.2 | - | 6, 7 | 1037 |