Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028947.1 | NP_083223.1 | 1 | 66 | ||
GenBank mRNA | AK016872.1 | - | 1 | 67 |