Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028948.2 | NP_083224.2 | 3 | 504 | ||
GenBank mRNA | AK016821.1 | - | 4 | 715 | ||
BC099500.1 | - | 3 | 422 | 30932292 | ||
BC125572.1 | - | 3 | 394 | 40130087 | ||
BC125574.1 | - | 3 | 394 | 40130089 |