Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028948.2 | NP_083224.2 | 2 | 313 | ||
GenBank mRNA | AK016821.1 | - | 3 | 524 | ||
AK162352.1 | - | 2 | 244 | |||
BC099500.1 | - | 2 | 231 | 30932292 | ||
BC125572.1 | - | 2 | 203 | 40130087 | ||
BC125574.1 | - | 2 | 203 | 40130089 | ||
CN835862.1 | - | 323 |