Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc74b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145435.1 | NP_001138907.1 | 8 | 1325 | ||
NM_029053.3 | NP_083329.2 | 9 | 1421 | |||
GenBank mRNA | AK132588.1 | - | 8 | 1323 | ||
AK143245.1 | - | 8 | 1298 | |||
AK161226.1 | - | 8 | 1323 |