Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029057.1 | NP_083333.1 | 12 | 3093 | ||
GenBank mRNA | AK044521.1 | - | 12 | 2900 | ||
AK049989.1 | - | 12 | 2926 | |||
AK173079.1 | - | 12 | 2856 | |||
BC059081.1 | - | 4 | 1601 | |||
BC072607.1 | - | 12 | 2919 | |||
BC079587.1 | - | 9 | 2402 | |||
BC138285.1 | - | 12 | 3300 | 8861304 | ||
BC138286.1 | - | 12 | 3300 | 8861305 |