Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930480E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177966.2 | NP_001171437.1 | 1 | 682 | ||
GenBank mRNA | BC116859.1 | - | 1 | 682 | 40126178 | |
BC116861.1 | - | 1 | 682 | 40126180 |