Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029152.1 | NP_083428.1 | 2 | 359 | ||
GenBank mRNA | AK015697.1 | - | 2 | 361 | ||
BC099564.1 | - | 2 | 359 | |||
BC119387.1 | - | 2 | 254 | |||
BC120662.1 | - | 2 | 254 |