Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029210.1 | NP_083486.1 | 13 | 3124 | ||
GenBank mRNA | AK173092.1 | - | 14 | 3042 |