Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm20l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029227.1 | NP_083503.1 | 3 | 257 | ||
GenBank mRNA | AK016101.1 | - | 3 | 242 | ||
BC048520.1 | - | 2 | 213 |