Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167691.1 | NP_001161163.1 | 1, 2 | 253 | ||
GenBank mRNA | AK016400.1 | - | 1, 2 | 253 |