Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029280.4 | NP_083556.2 | 3 | 502 | ||
GenBank mRNA | AK013063.1 | - | 3 | 485 | ||
AK133594.1 | - | 3 | 492 | |||
BC027547.1 | - | 3 | 435 | 1531973 | ||
CJ063020.1 | - | 3 | 492 |