Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019258.1 | NP_062131.1 | 2 | 377 | ||
GenBank mRNA | AF090692.1 | - | 2 | 377 |