Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029330.2 | NP_083606.2 | 4 | 2560 | ||
GenBank mRNA | AK045369.1 | - | 4 | 2561 |